missing translation for 'onlineSavingsMsg'
Learn More

Dicoumarol, MedChemExpress

Product Code. 16479116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479116 Supplier Medchem Express Supplier No. HYN0645

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Dicoumarol is an inhibitor of both NAD(P)H:quinone oxidoreductase 1 (NQO1) and PDK1 with IC50s of 0.37 and 19.42 μM, respectively.

Dicoumarol is an inhibitor of both NAD(P)H:quinone oxidoreductase 1 (NQO1) and PDK1 with IC50s of 0.37 and 19.42 μM, respectively.

Specifications

Chemical Name or Material Dicoumarol
CAS 66-76-2
Color White
Purity Grade Notes Research
Molecular Formula C19H12O6
Quantity 100 mg
Synonym Dicumarol
Solubility Information H2O : 25 mg/mL (74.34 mM; ultrasonic and adjust pH to 11 with NaOH) ∣DMSO : 3.67 mg/mL (10.91 mM; Need ultrasonic)
SMILES O=C1C(CC2=C(O)C3=CC=CC=C3OC2=O)=C(O)C4=CC=CC=C4O1
Molecular Weight (g/mol) 336.29
Formula Weight 336.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.