missing translation for 'onlineSavingsMsg'
Learn More

Dicofol (>85%), TRC

Product Code. 30384510
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384510 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384510 Supplier Toronto Research Chemicals Supplier No. TRCD43670010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dicofol
CAS 115-32-2
Molecular Formula C14 H9 Cl5 O
Quantity 10 g
Synonym Benzhydrol, 4,4'-dichloro-α-(trichloromethyl)- (8CI), 4-Chloro-α-(4-chlorophenyl)-α-(trichloromethyl)benzenemethanol, 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethanol, 1,1-Bis(p-chlorophenyl)-2,2,2-trichloroethanol, 2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethanol, 2,2,2-Trichloro-1,1-bis(p-chlorophenyl)ethanol, 2,2,2-Trichloro-1,1-di(4-chlorophenyl)ethanol, 4,4'-Dichloro-α-(trichloromethyl)benzhydrol, AFD 25, Acarin, Agravertin, CPCA, Chloretanol, Colonel-S, DTMC, Dacomain, Decofol, Dichlorokelthane, Dicofol, Dicofol kelthene, ENT 23,648, Ethanol, 2,2,2-trichloro-1,1-bis(4-chlorophenyl)-, FW 293, Keltane, Kelthane, Kelthane A, Kelthanethanol, Milbol, Mitigan, p,p'-Dicofol, p,p'-Kelthane
InChI Formula InChI=1S/C14H9Cl5O/c15-11-5-1-9(2-6-11)13(20,14(17,18)19)10-3-7-12(16)8-4-10/h1-8,20H
SMILES OC(c1ccc(Cl)cc1)(c2ccc(Cl)cc2)C(Cl)(Cl)Cl
IUPAC Name 2,2,2-trichloro-1,1-bis(4-chlorophenyl)ethanol
Molecular Weight (g/mol) 370.49
Formula Weight 367.9096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.