missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Sodium, MedChemExpress

Product Code. 16458887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458887 Supplier Medchem Express Supplier No. HY15037
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Diclofenac Sodium (GP 45840) is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac Sodium induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.

Diclofenac Sodium (GP 45840) is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac Sodium induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.

Specifications

Chemical Name or Material Diclofenac Sodium
Physical Form Solid
CAS 15307-79-6
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C14H10Cl2NNaO2
For Use With (Application) COVID-19-immunoregulation
Synonym GP 45840
Solubility Information H2O : 10 mg/mL (31.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O[Na])CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl
Molecular Weight (g/mol) 318.13
Formula Weight 318.13
Percent Purity 99.92%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.