missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Monobromo Sodium Salt Impurity, TRC

Product Code. 30381695
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381695 1 mg 1mg
30382658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381695 Supplier Toronto Research Chemicals Supplier No. TRCD4364901MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate
CAS 127792-45-4
Molecular Formula C14 H10 Br Cl N O2 . Na
Quantity 1 mg
Synonym Sodium 2-[2-[(2-Bromo-6-chlorophenyl)amino]phenyl]acetate, Diclofenac Sodium Imp. D (EP) as Sodium Salt
InChI Formula InChI=1S/C14H11BrClNO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1
SMILES [Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Br
IUPAC Name sodium;2-[2-(2-bromo-6-chloroanilino)phenyl]acetate
Molecular Weight (g/mol) 362.58
Formula Weight 360.9481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.