missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Diethylamine Salt, TRC

Product Code. 30385429
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385429 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385429 Supplier Toronto Research Chemicals Supplier No. TRCD436740250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac Diethylamine
CAS 78213-16-8
Purity Grade Notes HPLC
Molecular Formula C14 H11 Cl2 N O2 . C4 H11 N
Quantity 250 mg
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, compd. with N-ethylethanamine (1:1), Ethanamine, N-ethyl-, 2-[(2,6-dichlorophenyl)amino]benzeneacetate (9CI), Atomo Desinflamante, Diclofenac diethylamine, Diclofenac diethylamine salt, Diclofenac diethylammonium salt, Voltaren Emulgel, Voltaren Schmerzgel
InChI Formula InChI=1S/C14H11Cl2NO2.C4H11N/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;1-3-5-4-2/h1-7,17H,8H2,(H,18,19);5H,3-4H2,1-2H3
SMILES CCNCC.OC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-(2,6-dichloroanilino)phenyl]acetic acid;N-ethylethanamine
Molecular Weight (g/mol) 369.29
Formula Weight 368.1058
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.