missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclofenac-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Diclofenac-D4 |
| CAS | 153466-65-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 2H4 H7 Cl2 N O2 |
| Quantity | 1 mg |
| Synonym | Benzene-2,3,4,5-d4-acetic acid, 6-[(2,6-dichlorophenyl)amino]-, 6-[(2,6-Dichlorophenyl)amino]-benzen-2,3,4,5-d4-acetic acid, Diclofenac D4 acid (phenyl D4), Diclofenac (phenyl D4) |
| InChI Formula | InChI=1S/C14H11Cl2NO2/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19/h1-7,17H,8H2,(H,18,19)/i1D,2D,4D,7D |
| SMILES | [2H]c1c([2H])c([2H])c(Nc2c(Cl)cccc2Cl)c(CC(=O)O)c1[2H] |
| IUPAC Name | 2-[2,3,4,5-tetradeuterio-6-(2,6-dichloroanilino)phenyl]acetic acid |
| Molecular Weight (g/mol) | 300.17 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction