missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Acyl-Beta-D-glucuronide Allyl Ester, TRC

Product Code. 30380590
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380590 25 mg 25mg
30372804 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380590 Supplier Toronto Research Chemicals Supplier No. TRCD43648525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698358-10-0
Molecular Formula C23 H23 Cl2 N O8
Molecular Weight (g/mol) 512.34
Synonym Diclofenac Acyl-B-D-Glucuronide Allyl Ester
IUPAC Name prop-2-enyl (2S,3S,4S,5R,6S)-6-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylate
SMILES O[C@@H]1[C@@H](O)[C@H](OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl)O[C@@H]([C@H]1O)C(=O)OCC=C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H23Cl2NO8/c1-2-10-32-22(31)21-19(29)18(28)20(30)23(34-21)33-16(27)11-12-6-3-4-9-15(12)26-17-13(24)7-5-8-14(17)25/h2-9,18-21,23,26,28-30H,1,10-11H2/t18-,19-,20+,21-,23+/m0/s1
Formula Weight 511.0801
Chemical Name or Material Diclofenac Acyl-Beta-D-glucuronide Allyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.