missing translation for 'onlineSavingsMsg'
Learn More

Diclazuril, TRC

Product Code. 30376069
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376069 Supplier Toronto Research Chemicals Supplier No. TRCD4362001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclazuril
CAS 101831-37-2
Purity Grade Notes HPLC
Molecular Formula C17 H9 Cl3 N4 O2
Quantity 1 g
Synonym 2,6-Dichloro-α-(4-chlorophenyl)-4-(4,5-dihydro-3,5-dioxo-1,2,4-triazin-2(3H)-yl)benzeneacetonitrile, Clinacox, Diclazuril, Nuoqiu, P 64433, R 64433, Vecoxan
InChI Formula InChI=1S/C17H9Cl3N4O2/c18-10-3-1-9(2-4-10)12(7-21)16-13(19)5-11(6-14(16)20)24-17(26)23-15(25)8-22-24/h1-6,8,12H,(H,23,25,26)
SMILES Clc1ccc(cc1)C(C#N)c2c(Cl)cc(cc2Cl)N3N=CC(=O)NC3=O
IUPAC Name 2-(4-chlorophenyl)-2-[2,6-dichloro-4-(3,5-dioxo-1,2,4-triazin-2-yl)phenyl]acetonitrile
Molecular Weight (g/mol) 407.64
Formula Weight 405.9791
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.