missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclazuril, MedChemExpress
Description
Diclazuril (R-64433), a benzeneacetonitrile derivative, is a potent and orally active anticoccidial agent. Diclazuril can be used for the research of certain infectious and parasitic diseases, including coccidiosis, acute toxoplasmosis, equine protozoal pyoencephalitis (EPM) et.al.
Specifications
Specifications
| Chemical Name or Material | Diclazuril |
| Physical Form | Solid |
| CAS | 101831-37-2 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H9Cl3N4O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | R-64433 |
| Solubility Information | DMSO : 25 mg/mL (61.33 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?