missing translation for 'onlineSavingsMsg'
Learn More

Diclazuril 6-Carboxamide, TRC

Product Code. 30387062
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387062 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387062 Supplier Toronto Research Chemicals Supplier No. TRCD43621525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclazuril 6-Carboxamide
CAS 1797132-12-7
Molecular Formula C18 H10 Cl3 N5 O3
Quantity 25 mg
Synonym 1,2,4-Triazine-6-carboxamide, 2-[3,5-dichloro-4-[(4-chlorophenyl)cyanomethyl]phenyl]-2,3,4,5-tetrahydro-3,5-dioxo-, Diclazuril 6-Carboxamide, (RS)-2-[3,5-Dichloro-4-[(4-chlorophenyl)cyanomethyl]phenyl]-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carboxamide, 2-[3,5-Dichloro-4-[(RS)-(4-chlorophenyl)cyanomethyl]phenyl]-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carboxamide
InChI Formula InChI=1S/C18H10Cl3N5O3/c19-9-3-1-8(2-4-9)11(7-22)14-12(20)5-10(6-13(14)21)26-18(29)24-17(28)15(25-26)16(23)27/h1-6,11H,(H2,23,27)(H,24,28,29)
SMILES NC(=O)C1=NN(C(=O)NC1=O)c2cc(Cl)c(C(C#N)c3ccc(Cl)cc3)c(Cl)c2
IUPAC Name 2-[3,5-dichloro-4-[(4-chlorophenyl)-cyanomethyl]phenyl]-3,5-dioxo-1,2,4-triazine-6-carboxamide
Molecular Weight (g/mol) 450.66
Formula Weight 448.9849
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.