missing translation for 'onlineSavingsMsg'
Learn More

Dicholoromethane, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 15594055 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15594055 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594055 Supplier JT Baker Supplier No. 9410.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

A group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations.

Specifications

Melting Point -95.1°C
Color Colorless
Density At 25°C is 1.315 - 1.321g/ml
Quantity 2.5 L
Odor Characteristic
Packaging Glass
Residue after Evaporation max. 2ppm
Molecular Formula CH2Cl2
MDL Number MFCD00000881
Water max. 0.02 %
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
Boiling Point 40°C
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Percent Purity 99.8%
Chemical Name or Material Preservative (Amylene) about 20ppm
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.