missing translation for 'onlineSavingsMsg'
Learn More

Dichlorvos, TRC

Product Code. 30388061
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388061 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388061 Supplier Toronto Research Chemicals Supplier No. TRCD43595050MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlorvos
CAS 62-73-7
Molecular Formula C4 H7 Cl2 O4 P
Quantity 50 mg
Synonym Phosphoric acid, 2,2-dichlorovinyl dimethyl ester (6CI,8CI), Ethenol, 2,2-dichloro-, dimethyl phosphate (8CI), 2,2-Dichloroethenyl dimethyl phosphate, 2,2-Dichlorovinyl dimethyl phosphate, Alatox, Atgard, Atgard V, BAY-b 4986, Bibesol, Brevinyl, Brevinyl E 50, Canogard, Chlorvinphos, DDVP, DDVP (insecticide), Dash, Dedevap, Des, Des (phosphate), Dichlofos, Dichlorman, Dichlorovinphos, Dichlorovos, Dichlorvos, Dimethyl 2,2-dichloroethenyl phosphate, Dimethyl 2,2-dichlorovinyl phosphate, Dimethyl dichlorovinyl phosphate, Divipan, ENT 20738, Equigard, Equigel, Estrosel, Estrosol, Estrosol (pesticide), Fecama, Fekama, Ferag ID TM, Insectigas D, Lava 100EC, Mopari, NSC 6738, Nefrafos, Nerkol, Nogos, Nogos 500, Nogos 50EC, Nogos G, Novotox, Nuvan, Nuvan 100EC, Nuvan 500 EC, Nuvan 7, Nuvan 76EC, Nuvos, O,O-Dimethyl 2,2-dichlorovinyl phosphate, OKO, OMS 14, Panaplate, Phosvit, Phosvit 75, Prima U, SD 1750, Szklarniak, TAP 9VP, Task, Task (insecticide), Unifos, Unifos (pesticide), Unifos 50EC, Unitox, Vapona, Vapona Insecticide, Vaponite, Vinylofos, Vinylophos, Winylophos
InChI Formula InChI=1S/C4H7Cl2O4P/c1-8-11(7,9-2)10-3-4(5)6/h3H,1-2H3
SMILES COP(=O)(OC)OC=C(Cl)Cl
IUPAC Name 2,2-dichloroethenyl dimethyl phosphate
Molecular Weight (g/mol) 220.98
Formula Weight 219.9459
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.