Learn More
Dichlorotris(triphenylphosphine)ruthenium(II), Premion™, 99.95% (metals basis), Ru 10.2% min
Description
Dichlorotris(triphenylphosphine)ruthenium(II) serves as a precatalyst for the hydrogenation of alkenes, nitro compounds, ketones, carboxylic acids, synthesis of furans from allenyl sulfides and C-H Activation Catalyst in a Ruthenium/Lewis Acid System
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 132°C to 134°C |
| Color | Brown to Black |
| Quantity | 5 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene |
| Formula Weight | 958.86 |
| Percent Purity | 99.95% |
| Grade | Premion |
| Concentration or Composition (by Analyte or Components) | Ru 10.2% min. |
| Physical Form | Crystalline Powder |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.