Learn More
µ-Dichlorotetraethylene dirhodium(I), Acros Organics
Brand: Acros Organics 197590010
Code : 29
Additional Details : CAS Number : 12081-16-2 Weight : 0.05100kg
Packaging | Glass bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
12081-16-2 | |
388.92 | |
QPOCZCJMFQWGSP-UHFFFAOYSA-L | |
45357812 | |
C=C.C=C.C=C.C=C.[Cl-].[Cl-].[Rh].[Rh] |
C8H16Cl2Rh2 | |
MFCD00013206 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
ethene;rhodium;dichloride |
Specifications
µ-Dichlorotetraethylene dirhodium(I) | |
Glass bottle | |
1g | |
C8H16Cl2Rh2 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
QPOCZCJMFQWGSP-UHFFFAOYSA-L | |
ethene;rhodium;dichloride | |
45357812 | |
Crystalline Powder or Chunks |
Authentic | |
Rust | |
12081-16-2 | |
MFCD00013206 | |
Solubility in water: insoluble | |
C=C.C=C.C=C.C=C.[Cl-].[Cl-].[Rh].[Rh] | |
388.92 | |
388.92 |