missing translation for 'onlineSavingsMsg'
Learn More

Dichlororesorcinol, TRC

Product Code. 30380449
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380449 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380449 Supplier Toronto Research Chemicals Supplier No. TRCD43624525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlororesorcinol
CAS 137-19-9
Molecular Formula C6H4Cl2O2
Quantity 25 g
Synonym 1,5-Dichloro-2,4-dihydroxybenzene, 4,6-Dichloro-1,3-benzenediol, 4,6-Dichlororesorcin, 4,6-Dichlororesorcinol, NSC 60650
InChI Formula InChI=1S/C6H4Cl2O2/c7-3-1-4(8)6(10)2-5(3)9/h1-2,9-10H
SMILES Oc1cc(O)c(Cl)cc1Cl
IUPAC Name 4,6-dichlorobenzene-1,3-diol
Molecular Weight (g/mol) 179
Formula Weight 177.9588
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.