missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloroprop, TRC
Specifications
Specifications
| Chemical Name or Material | Dichlorprop |
| CAS | 120-36-5 |
| Molecular Formula | C9 H8 Cl2 O3 |
| Quantity | 250 mg |
| Synonym | Propionic acid, 2-(2,4-dichlorophenoxy)- (6CI,7CI,8CI), 2-(2,4-Dichlorophenoxy)propanoic acid, (±)-2,4-DP, (±)-2-(2,4-Dichlorophenoxy)propanoic acid, (±)-2-(2,4-Dichlorophenoxy)propionic acid, (±)-Dichlorprop, 2,4-DCPPA, 2,4-DP, 2,4-Dichlorophenoxypropionic acid, 2,4-Dichlorprop, 2-(2,4-DP), 2-(2,4-Dichlorophenoxy)propionic acid, AF 302, Canapur DP, Celatox DP, Cornox RK, DCP, Dichloroprop, Dichlorprop, Dichlorprop acid, Dicopur DP, Diprop, Hedonal DP, Herbizid DP, NSC 39624, U 46DP Fluid, α-(2,4-Dichlorophenoxy)propionic acid |
| InChI Formula | InChI=1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13) |
| SMILES | CC(Oc1ccc(Cl)cc1Cl)C(=O)O |
| IUPAC Name | 2-(2,4-dichlorophenoxy)propanoic acid |
| Molecular Weight (g/mol) | 235.06 |
| Formula Weight | 233.985 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?