missing translation for 'onlineSavingsMsg'
Learn More

Dichloroprop, TRC

Product Code. 30379940
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30379940

Brand: Toronto Research Chemicals TRCD4367451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlorprop
CAS 120-36-5
Molecular Formula C9 H8 Cl2 O3
Quantity 1 g
Synonym Propionic acid, 2-(2,4-dichlorophenoxy)- (6CI,7CI,8CI), 2-(2,4-Dichlorophenoxy)propanoic acid, (±)-2,4-DP, (±)-2-(2,4-Dichlorophenoxy)propanoic acid, (±)-2-(2,4-Dichlorophenoxy)propionic acid, (±)-Dichlorprop, 2,4-DCPPA, 2,4-DP, 2,4-Dichlorophenoxypropionic acid, 2,4-Dichlorprop, 2-(2,4-DP), 2-(2,4-Dichlorophenoxy)propionic acid, AF 302, Canapur DP, Celatox DP, Cornox RK, DCP, Dichloroprop, Dichlorprop, Dichlorprop acid, Dicopur DP, Diprop, Hedonal DP, Herbizid DP, NSC 39624, U 46DP Fluid, α-(2,4-Dichlorophenoxy)propionic acid
InChI Formula InChI=1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)
SMILES CC(Oc1ccc(Cl)cc1Cl)C(=O)O
IUPAC Name 2-(2,4-dichlorophenoxy)propanoic acid
Molecular Weight (g/mol) 235.06
Formula Weight 233.985
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.