missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylsilane, 95%

Product Code. 11945911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11945911 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11945911 Supplier Thermo Scientific Acros Supplier No. 432340250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1631-84-1 | C6H5Cl2Si | 176.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 1631-84-1
Color White to Yellow
Density 1.2040g/mL
Boiling Point 65.0°C to 66.0°C (10.0 mmHg)
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H5Cl2Si
Refractive Index 1.5245 to 1.5275 (20°C, 589nm)
MDL Number MFCD00039296
Quantity 25 mL
Specific Gravity 1.204
Synonym phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl
InChI Key XNAFLNBULDHNJS-UHFFFAOYSA-N
SMILES Cl[Si](Cl)C1=CC=CC=C1
IUPAC Name dichloro(phenyl)silyl
Molecular Weight (g/mol) 176.09
PubChem CID 6327324
Formula Weight 177.11
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Dichlorophenylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.