missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylphosphine, 97%

Product Code. 11494013
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494013 250 g 250g
11484013 50 g 50g
11474013 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494013 Supplier Thermo Scientific Alfa Aesar Supplier No. A10284.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 644-97-3 | C6H5Cl2P | 178.98 g/mol

Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylphosphine and its derivatives are used as an intermediate to make plasticizers, antioxidants, stabilizers, catalysts and high pressure lubricant additives and in lacquer formulatings.

Solubility
Reacts with water.

Notes
Air & moisture Sensitive. Incompatible with bases, oxidizing agents, water/ moisture &air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-97-3
Molecular Formula C6H5Cl2P
Molecular Weight (g/mol) 178.98
MDL Number MFCD00000528
InChI Key IMDXZWRLUZPMDH-UHFFFAOYSA-N
Synonym dichlorophenylphosphine, phenylphosphonous dichloride, phenyldichlorophosphine, benzenephosphonous dichloride, dichloro phenyl phosphine, phosphonous dichloride, phenyl, phenylphosphine dichloride, phenyl phosphorus dichloride, phenylphosphorus dichloride, phosphine, dichlorophenyl
PubChem CID 12573
IUPAC Name dichloro(phenyl)phosphane
SMILES ClP(Cl)C1=CC=CC=C1

Specifications

Melting Point -51°C
Density 1.319
Boiling Point 99°C to 101°C (11 mmHg)
Flash Point 102°C (215°F)
Odor Pungent
Refractive Index 1.598
Quantity 250 g
UN Number UN2798
Beilstein 508189
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 178.99
Percent Purity 97%
Chemical Name or Material Dichlorophenylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichlorophenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dichlorophenylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.