missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophen, MedChemExpress

Product Code. 16411279
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411279 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411279 Supplier Medchem Express Supplier No. HY12638
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dichlorophen (DDM) is an anticestodal agent. Dichlorophen is an antimicrobial agent shown to exert activity against cestodes, protozoa, fungi, and bacteria.

Dichlorophen (DDM) is an anticestodal agent. Dichlorophen is an antimicrobial agent shown to exert activity against cestodes, protozoa, fungi, and bacteria.

Specifications

Chemical Name or Material Dichlorophen
Physical Form Solid
CAS 97-23-4
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C13H10Cl2O2
For Use With (Application) COVID-19-immunoregulation
Synonym DDM
Solubility Information DMSO : ≥ 100 mg/mL (371.58 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C(Cl)C=C1CC2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 269.12
Formula Weight 269.12
Percent Purity 98.62%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.