missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 10076253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076253 2.5 kg Glass bottle 2.5kg
10488100 100 g Glass bottle 100g
10366690 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10076253 Supplier Thermo Scientific Acros Supplier No. 147430025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-70-9
MDL Number MFCD00000492
InChI Key YLJJAVFOBDSYAN-UHFFFAOYSA-N
Synonym dichloro methyl vinyl silane, methylvinyldichlorosilane, silane, dichloroethenylmethyl, vinylmethyldichlorosilane, methyldichlorovinylsilane, silane, dichloromethylvinyl, unii-b27y6f24py, ccris 2457, dichloromethylvinyl-silan, dichloromethylvinyl silane
PubChem CID 31299
IUPAC Name dichloro-ethenyl-methylsilane
SMILES C[Si](C=C)(Cl)Cl

Specifications

Color Colorless to Brown
Density 1.0800g/mL
Boiling Point 92.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H6Cl2Si
Linear Formula H2C=CHSi(CH3)Cl2
Refractive Index 1.4290 to 1.4310
Quantity 2.5 kg
Specific Gravity 1.08
Solubility Information Solubility in water: reacts.
Molecular Weight (g/mol) 141.07
Formula Weight 141.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichloromethylvinylsilane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethylvinylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.