missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylsilane, 97%, ACROS Organics™

Product Code. 10153190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10153190 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10153190 Supplier Thermo Scientific Acros Supplier No. 147395000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-54-7
Color Yellow to Brown
Density 1.1000g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula CH4Cl2Si
Linear Formula CH3SiHCl2
Refractive Index 1.3970 to 1.4000 (20°C, 589nm)
MDL Number MFCD00000494
Quantity 500 mL
Beilstein 04, I, 581
Synonym Methyldichlorosilane
Formula Weight 115.04
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Dichloromethylsilane, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.