missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylphenylsilane, 98%

Product Code. 10051550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051550 500 mL Glass bottle 500mL
10407730 10 mL Glass bottle 10mL
10031530 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051550 Supplier Thermo Scientific Acros Supplier No. 147385000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-74-6 | C7H8Cl2Si | 191.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-74-6
Molecular Formula C7H8Cl2Si
Molecular Weight (g/mol) 191.13
MDL Number MFCD00000490
InChI Key GNEPOXWQWFSSOU-UHFFFAOYSA-N
Synonym methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
PubChem CID 9006
IUPAC Name dichloro-methyl-phenylsilane
SMILES C[Si](Cl)(Cl)C1=CC=CC=C1

Specifications

Color Colorless
Density 1.1760g/mL
Boiling Point 205°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5Si(CH3)Cl2
Refractive Index 1.5180 to 1.52
Quantity 500 mL
Specific Gravity 1.176
Solubility Information Solubility in water: reacts with water
Formula Weight 191.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dichloromethylphenylsilane, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethylphenylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.