missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylene-dimethyliminium Chloride, TRC

Product Code. 30371736
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30371736 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30371736

Brand: Toronto Research Chemicals TRCD4379205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloromethylene-dimethyliminium Chloride
CAS 33842-02-3
Molecular Formula C3 H6 Cl2 N
Quantity 5 g
Synonym (Dichloromethylene)dimethylammonium chloride, Phosgeniminium chloride, Vilsmeier reagent
InChI Formula InChI=1S/C3H6Cl2N/c1-6(2)3(4)5/h1-2H3/q+1
SMILES C[N+](=C(Cl)Cl)C
IUPAC Name dichloromethylidene(dimethyl)azanium
Molecular Weight (g/mol) 126.992
Formula Weight 125.988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.