missing translation for 'onlineSavingsMsg'
Learn More

(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals

Product Code. 11415043
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33842-02-3
Molecular Formula C3H6Cl3N
Molecular Weight (g/mol) 162.44
MDL Number MFCD00011870
InChI Key NRNFKRFWZQQDMD-UHFFFAOYSA-M
Synonym dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
PubChem CID 2723945
IUPAC Name dichloromethylidene(dimethyl)azanium;chloride
SMILES [Cl-].CN(C)[C+](Cl)Cl

Specifications

Melting Point ∼185°C (decomposition)
Quantity 100 g
UN Number UN3261
Beilstein 4157987
Sensitivity Moisture sensitive
Formula Weight 162.45
Percent Purity 95%
Chemical Name or Material (Dichloromethylene)dimethylammonium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'