missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethyl-n-octylsilane, 98%

Product Code. 11389235
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389235 50 g 50g
11379235 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389235 Supplier Thermo Scientific Alfa Aesar Supplier No. L16565.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14799-93-0 | C9H20Cl2Si | 227.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14799-93-0
Molecular Formula C9H20Cl2Si
Molecular Weight (g/mol) 227.24
MDL Number MFCD00042606
InChI Key QHBMMABVNRSRHW-UHFFFAOYSA-N
Synonym n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
PubChem CID 84649
IUPAC Name dichloro-methyl-octylsilane
SMILES CCCCCCCC[Si](C)(Cl)Cl

Specifications

Boiling Point 224°C to 226°C
Quantity 50 g
UN Number UN2987
Beilstein 1740697
Flash Point 98°C (208°F)
Formula Weight 227.25
Percent Purity 98%
Chemical Name or Material Dichloromethyl-n-octylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.