missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethyl Methyl Ether, TRC

Product Code. 30380643
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380643 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380643 Supplier Toronto Research Chemicals Supplier No. TRCD43544025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloromethyl Methyl Ether
CAS 4885-02-3
Molecular Formula C2H4Cl2O
Quantity 25 g
Synonym 1,1-Dichlorodimethyl Ether, 1,1-Dichloromethyl Methyl Ether, Chloromyl, Dichloromethoxymethane, Dichloromethyl Methyl Ether, Methoxydichloromethane, Methyl alpha,alpha-dichloromethyl Ether, NSC 91480, alpha,alpha-Dichloromethyl Methyl Ether
InChI Formula InChI=1S/C2H4Cl2O/c1-5-2(3)4/h2H,1H3
SMILES COC(Cl)Cl
IUPAC Name dichloro(methoxy)methane
Molecular Weight (g/mol) 114.96
Formula Weight 113.9639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.