missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals

Catalog Number p-3757597
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4885-02-3
C2H4Cl2O
114.96
MFCD00000828
GRTGGSXWHGKRSB-UHFFFAOYSA-N
COC(Cl)Cl
This item is not returnable. View return policy
4885-02-3
C2H4Cl2O
114.96
MFCD00000828
GRTGGSXWHGKRSB-UHFFFAOYSA-N
COC(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 85.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2CHOCH3
Refractive Index 1.4295 to 1.4325
Quantity 10 g
Fieser 01,220; 02,120; 05,200; 06,171; 07,98; 08,154
Specific Gravity 1.27
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
IUPAC Name dichloro(methoxy)methane
PubChem CID 21004
Formula Weight 114.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 1-Dichlorodimethyl ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1-Dichlorodimethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • EUH018-In use, may form flammable/explosive vapour-air mixture.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'