missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene

Product Code. p-7049972
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11308377

Brand: Thermo Scientific Alfa Aesar 032440.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Dichloromethane is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -95°C
Density 1.325 g/mL
Boiling Point 39°C to 40°C
Quantity 1 L
Odor Sweet
Physical Form Liquid
Refractive Index 1.4244
UN Number UN1593
Beilstein 1730800
Merck Index 14,6063
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Formula Weight 84.93
Percent Purity ≥99.7%
Grade Spectrophotometric
Chemical Name or Material Dichloromethane, Stabilized with amylene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.