Learn More
Dichloromethane Reagent - British Pharmacopoeia, Fisherbrand™
Product Code. 30695000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Product Code. Quantity
30695000 1000 mL
1 options
Product Code. 30695000 Supplier Fisher Chemical Supplier No. BPR 105
Pharmacopoeia reagents for testing and monographs, produced under a GMP-compliant ISO 9001 quality management system.
    This Pharmacopoeia reagent is manufactured :
  • Using pharmacopoeia reagent quality components
  • Water meeting the relevant pharmacopoeia monograph requirement for Purified Water
  • Supplied with a Certificate of Analysis for Performance Test
  • Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO 17034
Hazard Statement: May be flammable or oxidizing; may cause severe skin burns and serious eye damage or irritation; may cause respiratory irritation, drowsiness or dizziness; may be toxic if swallowed, in contact with skin, or if inhaled; may cause cancer, genetic defects, reproductive toxicity or developmental harm; may be harmful to aquatic life with long-lasting effects; may be corrosive to metals.
Specifications
Quantity 1000 mL