missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, for HPLC, Unstabilised, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
Specifications
Specifications
| Melting Point | -97°C |
| Color | Colorless |
| Boiling Point | 39°C |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Formula Weight | 84.93g/mol |
Safety and Handling
Product Identifier
- Dichloromethane, unstabilized
- Warning
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction