missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane (DCM), Glass Distilled, Greenfield
Specifications
Specifications
| CAS | 75-09-2 |
| Quantity | 4x4L Case |
| Packaging | 4x4L Case |
| Boiling Point | 40°C |
| Grade | HPLC |
| Chemical Name or Material | Dichloromethane (DCM), Glass Distilled |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?