missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane (DCM), ACS, Greenfield
Click to view available options
:
4 x 4 L
Specifications
Specifications
| CAS | 75-09-2 |
| Boiling Point | 40°C |
| Quantity | 4x4L Case |
| Packaging | 4x4L Case |
| Grade | Reagent |
| Chemical Name or Material | Dichloromethane (DCM), ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction