missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D

Product Code. 16317731 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16317731 7.5 mL 7.50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16317731 Supplier Thermo Scientific Supplier No. 217430075

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1665-00-5 | CH2Cl2 | 86.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.3600 g/mL
Refractive Index 1.4218
Specific Gravity 1.36
Quantity 7.5 mL
CAS 1665-00-5
Melting Point -97.0°C
Boiling Point 40.0°C
Molecular Formula CH2Cl2
Linear Formula CD2Cl2
Beilstein 01, IV, 39
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in alchol, ether and dmf
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
SMILES [2H]C([2H])(Cl)Cl
IUPAC Name dichloro(²H₂)methane
Molecular Weight (g/mol) 86.94
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.