missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, 99.8 atom % D

Product Code. 16307731 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16307731 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 16307731 Supplier Thermo Scientific Supplier No. 214550100
  • ONLINE EXCLUSIVE PRICE
    £224.00
    £205.00 / 10mL
    Save £19.00
    8% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1665-00-5 | CH2Cl2 | 86.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.3600 g/mL
Refractive Index 1.4218
Specific Gravity 1.36
Quantity 10 mL
CAS 1665-00-5
Melting Point -97.0°C
Boiling Point 40.0°C
Molecular Formula CH2Cl2
Linear Formula CD2Cl2
Beilstein 01, IV, 39
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in alchol, ether and dmf
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
SMILES [2H]C([2H])(Cl)Cl
IUPAC Name dichloro(²H₂)methane
Molecular Weight (g/mol) 86.94
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane-d2, for NMR, 99.8 atom % D
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dichloromethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.