missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, CHROMASOLV™ AMD, stabilized, ≥99.8% (GC), Solstice

Product Code. 15684690
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15684690 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684690 Supplier Honeywell-Chromasolv Supplier No. 348976X1L
  • ONLINE EXCLUSIVE PRICE
    £296.00
    £254.68 / Pack of 6
    Save £41.32
    13% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CHROMASOLV™ AMD, stabilized,≥99.8% (GC)

  • Solvents designed for high-performance thin-layer chromatography
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -97°C (literature)
CAS 75-09-2
CAS Min % ≥99.0000%
CAS Max % ≥99.0000%
Density 1.325 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Packaging Glass Bottle
Molecular Formula CH2Cl2
Linear Formula CH2Cl2
MDL Number MFCD00000881
UN Number UN1593
Beilstein 1730800
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
Boiling Point 39°C to 41°C
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93
Percent Purity >99.8%
Grade High Purity
Chemical Name or Material Dichloromethane, CHROMASOLV™ AMD, stabilized, ≥99.8% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.