missing translation for 'onlineSavingsMsg'
Learn More

Solstice Dichloromethane CHROMASOLV™

Product Code. 15704358
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15704358 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15704358 Supplier Solstice Supplier No. 3485618L
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dichloromethane CHROMASOLV™ for HPLC, ≥99.8%, contains amylene as stabilizer.

  • Low UV profile by HPLC.

Specifications

pH Not applicable
Melting Point -97°C
CAS 75-09-2
CAS Min % ≥99.00%
Color Colorless
Density 1.32 to 1.33 g/cm3 (20°C)
Quantity 18 L
Flash Point Not applicable
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Molecular Formula CH2Cl2
Linear Formula CH2Cl2
Vapor Pressure 453 hPa (20°C)
MDL Number MFCD00000881
Viscosity 0.44 mPa.s (20°C)
UN Number UN1593
Beilstein 1730800
Concentration ≥99.80%
Synonym Methylene chloride
Boiling Point 39°C to 41°C (1.013 hPa)
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Percent Purity ≥99.80%
Grade HPLC
Chemical Name or Material Dichloromethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.