missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™

Product Code. 10616642 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616642 25 L Metal Can 25 L
10160292 1 L Amber Glass Bottle 1 L
10203810 1 L Plastic-coated Glass Bottle 1 L
10784941 2.5 L Amber Glass Bottle 2.5 L
10676062 2.5 L Plastic-coated Glass Bottle 2.5 L
10386272 5 L Plastic Canister 5 L
10459973 10 L Metal Can 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10616642 Supplier Fisher Chemical Supplier No. D/1852/25
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
Boiling Point 39°C
Formula Weight 84.93g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.