missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane Anhydrous stabilised with amylene extra dry, Solstice

Product Code. 15867361
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867361 1 L 1 L
15857361 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15867361 Supplier Honeywell Riedel-de Haën Supplier No. 662041L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contain less that 50 ppm water. Anhydrous Solvents are packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity.

  • These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
  • Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
  • All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
  • The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
  • This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Density 1.32 to 1.33 g/cm3 (20°C)
Flash Point Not applicable
Packaging Septum Sealed Bottle
Quantity 1 L
UN Number 1593
Percent Purity <0.0010% H2O
Chemical Name or Material Dichloromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.