missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS, 99.5+%, stab. with amylene

Product Code. 11383488
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
4 x 1 L
Unit Size:
1 L
2.5 L
4 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383488 2.5 L 2.5 L
11480211 500 mL 500mL
11393488 1 L 1 L
11303498 4 x 1 L 4 L
11373488 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11383488 Supplier Thermo Scientific Alfa Aesar Supplier No. 039116.K4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02% | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Appearance Colorless liquid
Density 1.325
Quantity 2.5 L
Ignition Point 605°C (1121°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.4244
Vapor Pressure 475 hPa (356mm Hg) at 20°C
Beilstein 1730800
Boiling Point 39°C to 40°C
Solubility Information Miscible in ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform.
Formula Weight 84.93
Grade ACS
Chemical Name or Material Dichloromethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.