missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
| Density | 1.3250g/mL |
| Quantity | 10 L |
| Packaging | Metal Drum |
| Physical Form | Liquid |
| Residue after Evaporation | 0.001% max. |
| Assay Percent Range | 99.8+% |
| Linear Formula | CH2Cl2 |
| Water | 0.02% max. (Coulometric) |
| Beilstein | 01, 60 |
| Fieser | 01,676; 02,273; 04,337; 07,239; 13,106 |
| Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?