missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.6%, ACS reagent, stabilized with amylene

Product Code. 10532904 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
This item is not returnable. View return policy

Product Code. 10532904

Brand: Thermo Scientific Acros 406920250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Decomposition Information >120°C
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Quantity 25 L
Packaging Metal Drum
Physical Form Liquid
Assay Percent Range 99.6%
Residue after Evaporation 0.002% max.
Specific Gravity 1.325
Titratable Acid 0.0003 meq/g max.
Linear Formula CH2Cl2
Vapor Pressure Vapor Pressure: 475hPa at 20°C
Viscosity 0.43 mPa.s (20°C)
Beilstein 01, 60
Water 0.02% max. (Coulometric)
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.6%
Grade ACS Reagent
Chemical Name or Material Dichloromethane, Stabilized with Amylene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.