missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ≥99.5% (GC), Honeywell™ Riedel-de Haën™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
This item is not returnable. View return policy
75-09-2
CH2Cl2
84.927
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
C(Cl)Cl
UN Number UN1593
Beilstein 1730800
Formula Weight 84.93g/mol
Percent Purity ≥99.5% (GC)
Density 1.325g/mL at 25°C
Color Colorless
Quantity 250mL
Explosion Limit 0.22
Ignition Point 1223°C
Odor Odorless
Packaging Glass bottle
Physical State Liquid
Vapor Density 2.9 (vs air)
Vapor Pressure 6.83mmHg (20°C), 24.45mmHg (55°C)
Chemical Name or Material Dichloromethane
Show More Show Less