missing translation for 'onlineSavingsMsg'
Learn More

Dichloroisocyanuric Acid Sodium Salt, TRC

Product Code. 30385214
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385214 Supplier Toronto Research Chemicals Supplier No. TRCD43487525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloroisocyanuric Acid Sodium Salt
CAS 2893-78-9
Molecular Formula C3Cl2N3NaO3
Quantity 25 g
Synonym Dichloroisocyanuric Acid Sodium, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3-dichloro-, sodium salt (9CI), s-Triazine-2,4,6(1H,3H,5H)-trione, 1,3-dichloro-, sodium salt (8CI), 3,5-Dichloro-2-hydroxy-4,6-s-triazinedione sodium salt, ACL 56, ACL 60, Actisan, Actisept, Aviclor, Basolan DC, CDB 63, CDB Clearon, Clearon, Clearon CDB, Clearon CDB 56, Crente, DICD-G, Dichloro-s-triazinetrione sodium salt, Dichloroisocyanuric acid sodium salt, Dichlosia, Dikonit, Fi Clor 60S, Fi Clor Clearon, Fichlor, Haz-Tab, Haz-Tabs, Hi-Lite 60C, Hi-Lite 60G, Hi-Lite G 60GW, Hidrosan, Histar G, Izosan G, Javel Solid, Jundujing, Khlorapin, Medicarine, Neochlor 55, Neochlor 55G, Neochlor 60G, Neochlor 60M, Neochlor 60MG, Neochlor 60P, Oniachlor 60, Oxidan DCN/WSG, Presept, Prisept, SDIC 60P, SDIC-MG, Simpla, Sodium dichlorisocyanurate, Sodium dichlorocyanurate, Sodium dichloroisocyanurate, Sodium dicycloroisocyanurate, Sumaveg, Surchlor GR 60, Vac Attack, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1,3-dichloro-, sodium salt (9CI)s-Triazine-2,4,6(1H,3H,5H)-trione, 1,3-dichloro-, sodium salt (8CI)3,5-Dichloro-2-hydroxy-4,6-s-triazinedione sodium saltACL 56, ACL 60, Actisan, Actisept, Aviclor, Basolan DC, CDB 63, CDB Clearon, Clearon, Clearon CDB, Clearon CDB 56, Crente, DICD-G, Dichloro-s-triazinetrione Sodium Salt, Dichloroisocyanuric acid Sodium SaltDichlosia, Dikonit, Fi Clor 60S, Fi Clor Clearon, Fichlor, Haz-Tab, Haz-Tabs, Hi-Lite 60C, Hi-Lite 60G, Hi-Lite G 60GW, Hidrosan, Izosan G, Javel Solid, Jundujing, Khlorapin, Medicarine, Neochlor 55, Neochlor 55G, Neochlor 60G, Neochlor 60M, Neochlor 60MG, Neochlor 60P, Oniachlor 60, Oxidan DCN/WSG, Presept, PriseptSDIC 60P, SDIC-MG, Simpla, Sodium Dichlorisocyanurate, Sodium Dichlorocyanurate, Sodium Dichloroisocyanurate, Sumaveg, Surchlor GR 60, Vac Attack
InChI Formula InChI=1S/C3HCl2N3O3.Na/c4-7-1(9)6-2(10)8(5)3(7)11;/h(H,6,9,10);/q;+1/p-1
SMILES [Na+].ClN1C(=O)[N-]C(=O)N(Cl)C1=O
IUPAC Name sodium;1,3-dichloro-1,3-diaza-5-azanidacyclohexane-2,4,6-trione
Molecular Weight (g/mol) 219.95
Formula Weight 218.9214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.