missing translation for 'onlineSavingsMsg'
Learn More

Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number 15274674
Click to view available options
Quantity:
25 g
500 g
2.5 kg
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]
2893-78-9
C3HCl2N3NaO3
220.949
MFCD00006036
UNWRHVZXVVTASG-UHFFFAOYSA-N
sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
86657659
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY
Color White
pH 6 to 7 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 2.5 kg
Formula Weight 219.95
Percent Purity ≥97.5% (on dry substance)
Physical Form Crystals or Granules
Chemical Name or Material Dichloroisocyanuric acid sodium salt, 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'