missing translation for 'onlineSavingsMsg'
Learn More

Dichloroisocyanuric acid sodium salt, 97% (dry wt.)

Product Code. p-7027052
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11314519

Brand: Thermo Scientific Alfa Aesar B23504.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2893-78-9 | C3HCl2N3NaO3 | 220.949 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2893-78-9
Molecular Formula C3HCl2N3NaO3
Molecular Weight (g/mol) 220.949
MDL Number MFCD00006036
InChI Key UNWRHVZXVVTASG-UHFFFAOYSA-N
Synonym sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
PubChem CID 86657659
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Specifications

Melting Point ∼250°C (decomposition)
Odor Chlorine-like
Quantity 25 g
UN Number UN2465
Beilstein 4056155
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 219.95
Percent Purity 97%
Assay (dry wt.), water <3%
Chemical Name or Material Dichloroisocyanuric acid sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.