missing translation for 'onlineSavingsMsg'
Learn More

Dichloroiodomethane, TRC

Product Code. 17943605
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943605 50 mg 50mg
17974045 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17943605 Supplier Toronto Research Chemicals Supplier No. TRCD43468050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-04-7
Molecular Formula C H Cl2 I
Molecular Weight (g/mol) 210.83
Synonym Dichloroiodomethane, Methane, dichloroiodo- (6CI,7CI,8CI,9CI)
IUPAC Name dichloro(iodo)methane
SMILES ClC(Cl)I

Specifications

Quantity 50 mg
InChI Formula InChI=1S/CHCl2I/c2-1(3)4/h1H
Formula Weight 209.85
Chemical Name or Material Dichloroiodomethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.