missing translation for 'onlineSavingsMsg'
Learn More

Dichloroimipramine Hydrochloride, TRC

Product Code. 30371122
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371122 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371122 Supplier Toronto Research Chemicals Supplier No. TRCD43484550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride
CAS 115189-28-1
Molecular Formula C19 H22 Cl2 N2 . Cl H
Quantity 50 mg
Synonym 3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Clomipramine Hydrochloride Imp. D (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 3,7-dichloro-10,11-dihydro-N,N-dimethyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 3,7-dichloro-5-(3-dimethylaminopropyl)-10,11-dihydro-, hydrochloride (6CI)
InChI Formula InChI=1S/C19H22Cl2N2.ClH/c1-22(2)10-3-11-23-18-12-16(20)8-6-14(18)4-5-15-7-9-17(21)13-19(15)23;/h6-9,12-13H,3-5,10-11H2,1-2H3;1H
SMILES Cl.CN(C)CCCN1c2cc(Cl)ccc2CCc3ccc(Cl)cc13
IUPAC Name 3-(2,9-dichloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 385.76
Formula Weight 384.0927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.