missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilicon, TRC

Product Code. 30137861
Change view
Click to view available options
Quantity:
2.5 g
10 g
100 g
Unit Size:
100g
10g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137861 10 g 10g
30137625 100 g 100g
30011146 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137861 Supplier Toronto Research Chemicals Supplier No. TRCD43447510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-78-5
Molecular Formula C2 H6 Cl2 Si
Molecular Weight (g/mol) 129.06
Synonym DMDCS, Dimethyldichlorsilane
IUPAC Name dichloro(dimethyl)silane
SMILES C[Si](C)(Cl)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3
Formula Weight 127.9616
Chemical Name or Material Dimethyldichlorosilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.