missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorodiethylsilane, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -96.0°C |
| Density | 1.0500g/mL |
| Boiling Point | 128.0°C to 129.0°C |
| Flash Point | 28°C |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (CH3CH2)2Si(Cl)2 |
| Quantity | 5 g |
| Specific Gravity | 1.05 |
| Solubility Information | Solubility in water: reacts |
| Show More |
Safety and Handling
Product Identifier
- Dichlorodiethylsilane
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction